missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyrazolo[1,5-a]pyrimidine, 97%

Product Code. 15466727
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466727 1 g 1g
15476727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466727 Supplier Thermo Scientific Alfa Aesar Supplier No. H33855.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 705263-10-1 | C6H4BrN3 | 198.02 g/mol

6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705263-10-1
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.02
MDL Number MFCD09832894
InChI Key VDHTXLUCUNPVLO-UHFFFAOYSA-N
Synonym 6-bromopyrazolo 1,5-a pyrimidine, 6-bromo-pyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 6-bromo, pubchem15248, acmc-209of0
PubChem CID 22236701
IUPAC Name 6-bromopyrazolo[1,5-a]pyrimidine
SMILES BrC1=CN2N=CC=C2N=C1

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 198.02
Percent Purity 97%
Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine
compliance-icons
Product Identifier
  • 6-Bromopyrazolo[1,5-a]pyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 6-Bromopyrazolo[1,5-a]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.