missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyrazolo[1,5-a]pyrimidine, 97%

Product Code. 15476727
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15476727

Brand: Thermo Scientific Alfa Aesar H33855.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 705263-10-1 | C6H4BrN3 | 198.02 g/mol

6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705263-10-1
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.02
MDL Number MFCD09832894
InChI Key VDHTXLUCUNPVLO-UHFFFAOYSA-N
Synonym 6-bromopyrazolo 1,5-a pyrimidine, 6-bromo-pyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 6-bromo, pubchem15248, acmc-209of0
PubChem CID 22236701
IUPAC Name 6-bromopyrazolo[1,5-a]pyrimidine
SMILES BrC1=CN2N=CC=C2N=C1

Specifications

Melting Point 122°C to 124°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 198.02
Percent Purity 97%
Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.