missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-2-carbaldehyde, ≥95%, Thermo Scientific™

Product Code. 10197052
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10197052 1 g 1g
10052174 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197052 Supplier Thermo Scientific Maybridge Supplier No. MO07535DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 34160-40-2
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD02683546
InChI Key QWFHFNGMCPMOCD-UHFFFAOYSA-N
Synonym 6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
PubChem CID 2757009
IUPAC Name 6-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Br)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 6-Bromopyridine-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.