missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinoline, 97%

Product Code. 15491258
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15491258

Brand: Thermo Scientific Alfa Aesar H55111.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5332-25-2 | C9H6BrN | 208.058 g/mol

6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-25-2
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00024023
InChI Key IFIHYLCUKYCKRH-UHFFFAOYSA-N
Synonym quinoline, 6-bromo, 6-bromo-quinoline, 6-bromo quinoline, 6-bromchinolin, 6-bromchinolin;, 6-br-quinoline, 6-bromooquinoline, acmc-20aits, pubchem2374, 6-bromoquinoline
PubChem CID 79243
IUPAC Name 6-bromoquinoline
SMILES C1=CC2=C(C=CC(=C2)Br)N=C1

Specifications

Melting Point 24°C
Density 1.538
Flash Point >110°C (230°F)
Refractive Index 1.663
Quantity 5 g
Solubility Information Soluble in acetone,acetonitrile,dichloromethane,ethyl acetate and THF.
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 6-Bromoquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.