missing translation for 'onlineSavingsMsg'
Learn More

6-Bromothiochroman-4-one, 95%

Product Code. 15444617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444617 5 g 5g
15434617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444617 Supplier Thermo Scientific Alfa Aesar Supplier No. H31968.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13735-13-2 | C9H7BrOS | 243.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13735-13-2
Molecular Formula C9H7BrOS
Molecular Weight (g/mol) 243.118
MDL Number MFCD05664869
InChI Key SBNPYDJTVGRDAA-UHFFFAOYSA-N
Synonym 6-bromothiochroman-4-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro-1-benzothiopyran-4-one, 6-bromo-3,4-dihydro-2h-1-benzothiin-4-one, 6-bromo-thiochroman-4-one, 6-bromo-2h-1-benzothiopyran-4 3h-one, 6-bromo-2,3-dihydro-4h-1-benzothiopyran4-one, 2,3-dihydro-6-bromo-4h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one, 6-bromo-2,3-dihydro
PubChem CID 13953736
IUPAC Name 6-bromo-2,3-dihydrothiochromen-4-one
SMILES C1CSC2=C(C1=O)C=C(C=C2)Br

Specifications

Melting Point 66°C to 69°C
Quantity 5 g
Formula Weight 243.12
Percent Purity 95%
Chemical Name or Material 6-Bromothiochroman-4-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.