missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-1,3-benzothiazol-2-amine, 97%, Thermo Scientific™

Product Code. 10471674
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10471674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10471674 Supplier Thermo Scientific Maybridge Supplier No. BTB04800DA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 95-24-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5ClN2S
MDL Number MFCD00053557
Quantity 1 g
Synonym 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
InChI Key VMNXKIDUTPOHPO-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1Cl)SC(=N2)N
IUPAC Name 6-chloro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol) 184.641
PubChem CID 7226
Formula Weight 184.65
Percent Purity 97%
Chemical Name or Material 6-Chloro-1, 3-benzothiazol-2-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.