missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1-hexanol, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-1-hexanol |
| CAS | 2009-83-8 |
| Molecular Formula | C6H13ClO |
| Quantity | 100 g |
| Synonym | 1-Chloro-6-hexanol, 1-Chloro-6-hydroxyhexane, 6-Chlorohexanol, Hexamethylene Chlorohydrin, NSC 3700, ω-Chlorohexanol |
| InChI Formula | InChI=1S/C6H13ClO/c7-5-3-1-2-4-6-8/h8H,1-6H2 |
| SMILES | OCCCCCCCl |
| IUPAC Name | 6-chlorohexan-1-ol |
| Molecular Weight (g/mol) | 136.62 |
| Formula Weight | 136.0655 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction