missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole, 95+%, Thermo Scientific™

Product Code. 10571243
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571243 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571243 Supplier Thermo Scientific Maybridge Supplier No. AW00326DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 348134-09-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Molecular Formula C12H14ClN3S
MDL Number MFCD00661816
Quantity 1 g
Synonym 6-chloro-2-1,4-diazepan-1-yl-1,3-benzothiazole, maybridge1_000355, 6-chloro-2-1,4-diazepane-1-yl-1,3-benzothiazole, 6-chloro-2-1,4 diazepan-1-yl-benzothiazole, 6-chloro-2-1.4 diazepan-1-yl-benzothiazole, 6-chloro-2-1,4-diazepan-1-yl benzo d thiazole
InChI Key UZLJYDCBJULBQU-UHFFFAOYSA-N
SMILES C1CNCCN(C1)C2=NC3=C(S2)C=C(C=C3)Cl
IUPAC Name 6-chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole
Molecular Weight (g/mol) 267.775
PubChem CID 2795675
Formula Weight 267.78
Percent Purity 95%
Chemical Name or Material 6-Chloro-2-(1, 4-diazepan-1-yl)-1, 3-benzothiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.