missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-(chloromethyl)-4-phenyl-quinazoline, TRC

Product Code. 30350345
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350345 100 mg 100mg
30354352 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350345 Supplier Toronto Research Chemicals Supplier No. TRCC293300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5958-24-7
Molecular Formula C15 H10 Cl2 N2 O
Molecular Weight (g/mol) 305.16
Synonym 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide, Chlordiazepoxide Hydrochloride Imp. B (EP)
IUPAC Name 6-chloro-2-(chloromethyl)-3-oxido-4-phenylquinazolin-3-ium
SMILES [O-][n+]1c(CCl)nc2ccc(Cl)cc2c1c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H10Cl2N2O/c16-9-14-18-13-7-6-11(17)8-12(13)15(19(14)20)10-4-2-1-3-5-10/h1-8H,9H2
Formula Weight 304.017
Chemical Name or Material 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.