missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2H-chromene-3-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10304342
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10304342 10 g 10g
10140432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304342 Supplier Thermo Scientific Maybridge Supplier No. RDR03724EA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-54-6
Molecular Formula C10H6Cl2O2
Molecular Weight (g/mol) 229.056
MDL Number MFCD02091502
InChI Key HGFGWTNRUGRBNQ-UHFFFAOYSA-N
Synonym 6-chloro-2h-1-benzopyran-3-carbonyl chloride, 2h-1-benzopyran-3-carbonylchloride, 6-chloro, 6-chloro-2h-1-benzopyran-3-carbonylchloride, 6-chloranyl-2h-chromene-3-carbonyl chloride
PubChem CID 2735716
IUPAC Name 6-chloro-2H-chromene-3-carbonyl chloride
SMILES C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 229.06
Percent Purity 95%
Chemical Name or Material 6-Chloro-2H-chromene-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.