missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-3-(hydroxymethyl)pyridine, 98%, Thermo Scientific™

Product Code. 11986311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986311 1 g 1g
11916321 5 g 5g
11906321 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11986311 Supplier Thermo Scientific Acros Supplier No. 358090010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 21543-49-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
InChI Key GOXYBEXWMJZLJB-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol
PubChem CID 177161
IUPAC Name (6-chloropyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.7°C to 48.5°C
Color White to Brown
Density 1.2800g/mL
Boiling Point 190.0°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.28
Formula Weight 143.57
Percent Purity 98%
Physical Form Powder or Low Melting Solid
Chemical Name or Material 6-chloro-3-(hydroxymethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-hydroxymethylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.