missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97+%, Thermo Scientific™

Product Code. 11467214
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467214 5 g 5g
11457214 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467214 Supplier Thermo Scientific Alfa Aesar Supplier No. B21597.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18706-21-3
Molecular Formula C10H5ClF3NO
Molecular Weight (g/mol) 247.601
MDL Number MFCD00276583
InChI Key MGEMMMJZAWYWNI-UHFFFAOYSA-N
Synonym 6-chloro-4-hydroxy-2-trifluoromethyl quinoline, 6-chloro-2-trifluoromethyl quinolin-4-ol, 6-chloro-2-trifluoromethyl-1h-quinolin-4-one, 6-chloro-2-trifluoromethyl-quinolin-4-ol, 6-chloro-2-trifluoromethyl quinolin-4 1h-one, 6-chloro-2-trifluoromethylquinolin-4-ol, acmc-1boyc, 6-chloro-2-trifluoromethyl quinoline-4-ol, 2-trifluoromethyl-4-hydroxy-6-chloroquinoline
PubChem CID 2736609
IUPAC Name 6-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
SMILES C1=CC2=C(C=C1Cl)C(=O)C=C(N2)C(F)(F)F

Specifications

Melting Point 278°C to 280°C
Quantity 5 g
Formula Weight 247.61
Percent Purity ≥97%
Chemical Name or Material 6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.