missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-methyl-4H-chromen-4-one, 98%, BLD Pharm™

Product Code. 31422382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31422382 250 mg
31417295 5 g
31418976 100 mg
31420809 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31422382 Supplier BLD Pharm Supplier No. BD108878250mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifikationer

CAS 67029-84-9
Molecular Formula C10H7ClO2
MDL Number MFCD00239401
Quantity 250 mg
SMILES O=C1C=COC2=C1C=C(Cl)C(C)=C2
Molecular Weight (g/mol) 194.61
Percent Purity 98%
Chemical Name or Material 6-Chloro-7-methyl-4H-chromen-4-one
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.