missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-imidazo[2,1-b]thiazole-5-sulfonic Acid Amide, TRC

Product Code. 30354810
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354810 100 mg -
30366258 10 mg 10mg
30363395 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354810 Supplier Toronto Research Chemicals Supplier No. TRCC383335100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112582-89-5
Molecular Formula C5H4N3O2S2Cl
Molecular Weight (g/mol) 237.69
IUPAC Name 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonamide
SMILES NS(=O)(=O)c1c(Cl)nc2sccn12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H4ClN3O2S2/c6-3-4(13(7,10)11)9-1-2-12-5(9)8-3/h1-2H,(H2,7,10,11)
Formula Weight 236.9433
Chemical Name or Material 6-Chloro-imidazo[2,1-b]thiazole-5-sulfonic Acid Amide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.