missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorochroman-4-one, 99%

Product Code. 11467187
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467187 25 g 25g
11447187 1 g 1g
11457187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467187 Supplier Thermo Scientific Alfa Aesar Supplier No. A14975.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 37674-72-9 | C9H7ClO2 | 182.603 g/mol

6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

Solubility
Soluble in solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 37674-72-9
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00024070
InChI Key LLTDYHFVIVSQPJ-UHFFFAOYSA-N
Synonym 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
PubChem CID 95273
IUPAC Name 6-chloro-2,3-dihydrochromen-4-one
SMILES C1COC2=C(C1=O)C=C(C=C2)Cl

Specifica

Melting Point 101°C to 102°C
Quantity 25 g
Beilstein 124218
Solubility Information Soluble in solvents.
Formula Weight 182.61
Percent Purity 99%
Chemical Name or Material 6-Chlorochroman-4-one

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.