missing translation for 'onlineSavingsMsg'
Learn More

6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10279082
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10279082 250 mg 250mg
10520113 1 g 1g
10332115 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279082 Supplier Thermo Scientific Maybridge Supplier No. MO00885CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14714-24-0
Molecular Formula C7H4ClN3O2
Molecular Weight (g/mol) 197.578
MDL Number MFCD04974055
InChI Key HTUZJYQXKFDVLT-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-b pyridazine-2-carboxylic acid, 2-carboxy-6-chloroimidazo 1,2-b pyridazine, 6-chloro-imidazo 1,2-b pyridazine-2-carboxylic acid, pubchem23037, 6-chloroimidazo 1,2-b pyridazine-2-carboxylicacid, 6-chloro-2-imidazo 1,2-b pyridazinecarboxylic acid, 6-chloranylimidazo 1,2-b pyridazine-2-carboxylic acid, imidazo 1,2-b pyridazine-2-carboxylicacid, 6-chloro, 6-chloro-4-hydroimidazo 1,2-e pyridazine-2-carboxylic acid, imidazo 1,2-b pyridazine-2-carboxylic acid, 6-chloro
PubChem CID 2794637
IUPAC Name 6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid
SMILES C1=CC(=NN2C1=NC(=C2)C(=O)O)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 197.58
Percent Purity 90%
Chemical Name or Material 6-Chloroimidazo[1, 2-b]pyridazine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.