missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole, 99%

Product Code. 11450048
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450048 1 g 1g
11460048 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11450048 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor B23650.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17422-33-2 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17422-33-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005681
InChI Key YTYIMDRWPTUAHP-UHFFFAOYSA-N
Synonym 6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
PubChem CID 87111
ChEBI CHEBI:80918
IUPAC Name 6-chloro-1H-indole
SMILES ClC1=CC=C2C=CNC2=C1

Specifications

Melting Point 87°C to 90°C
Quantity 1 g
Beilstein 112345
Formula Weight 151.6
Percent Purity 99%
Chemical Name or Material 6-Chloroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.