missing translation for 'onlineSavingsMsg'
Learn More

6-chloronicotinamide, 98%, Thermo Scientific™

Product Code. 11986281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11986281 1 g 1g
11996281 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986281 Supplier Thermo Scientific Acros Supplier No. 109560010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6271-78-9
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.57
MDL Number MFCD00006242
InChI Key ZIJAZUBWHAZHPL-UHFFFAOYSA-N
Synonym 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
PubChem CID 80456
IUPAC Name 6-chloropyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(Cl)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204.4°C to 207.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Cl(C5H3N)CONH2
Quantity 1 g
Beilstein 22,44
Formula Weight 156.57
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 6-Chloronicotinamide
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloronicotinamide, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.