missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorooxindole, 98%

Product Code. 11434217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434217 100 g 100g
11414217 5 g 5g
11424217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434217 Supplier Thermo Scientific Alfa Aesar Supplier No. A19674.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 56341-37-8 | C8H6ClNO | 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56341-37-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00209962
InChI Key CENVPIZOTHULGJ-UHFFFAOYSA-N
Synonym 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
PubChem CID 736344
IUPAC Name 6-chloro-1,3-dihydroindol-2-one
SMILES C1C2=C(C=C(C=C2)Cl)NC1=O

Specifications

Melting Point 195°C to 199°C
Quantity 100 g
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 6-Chlorooxindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.