missing translation for 'onlineSavingsMsg'
Learn More

6-chlorooxindole, 98%, Thermo Scientific™

Product Code. 10041644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10041644 25 g Glass bottle 25g
10612445 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041644 Supplier Thermo Scientific Acros Supplier No. 399440250
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56341-37-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00209962
InChI Key CENVPIZOTHULGJ-UHFFFAOYSA-N
Synonym 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
PubChem CID 736344
IUPAC Name 6-chloro-1,3-dihydroindol-2-one
SMILES C1C2=C(C=C(C=C2)Cl)NC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195°C to 199°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 167.59
Percent Purity 98%
Chemical Name or Material 6-chlorooxindole
compliance-icons
Product Identifier
  • 6-Chlorooxindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.