missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine 2'-deoxyriboside, 97%

Product Code. 15476619
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476619 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476619 Supplier Thermo Scientific Alfa Aesar Supplier No. J64871.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4594-45-0 | C10H11ClN4O3 | 270.673 g/mol

6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

Notes
Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4594-45-0
Molecular Formula C10H11ClN4O3
Molecular Weight (g/mol) 270.673
MDL Number MFCD00083282
InChI Key PGEULCIODBNODW-FSDSQADBSA-N
Synonym 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine
PubChem CID 92132234
IUPAC Name (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O

Specifications

Color White
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 270.67
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Chloropurine 2'-deoxyriboside
compliance-icons
Product Identifier
  • 6-Chloropurine 2'-deoxyriboside
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contain: 6-Chloropurine 2-deoxyriboside

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.