missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine-9-(2,3-isopropylidene-Beta-D-ribofuranoside), TRC

Product Code. 30361456
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30361456 5 g
30360460 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361456 Supplier Toronto Research Chemicals Supplier No. TRCC3798855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39824-26-5
Molecular Formula C13 H15 Cl N4 O4
Molecular Weight (g/mol) 326.74
IUPAC Name [(3aR,4R,6R,6aR)-4-(6-chloropurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)n3cnc4c(Cl)ncnc34

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H15ClN4O4/c1-13(2)21-8-6(3-19)20-12(9(8)22-13)18-5-17-7-10(14)15-4-16-11(7)18/h4-6,8-9,12,19H,3H2,1-2H3/t6-,8-,9-,12-/m1/s1
Formula Weight 326.0782
Chemical Name or Material 6-Chloropurine-9-(2,3-isopropylidene-beta-D-ribofuranoside)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.