missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine-9-beta-D-ribofuranoside, TRC

Product Code. 30368733
Change view
Click to view available options
Quantity:
1 g
10 g
500 g
Unit Size:
10g
1g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368733 10 g 10g
30364425 1 g 1g
30356572 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368733 Supplier Toronto Research Chemicals Supplier No. TRCC37990010G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5399-87-1
Molecular Formula C10 H11 Cl N4 O4
Molecular Weight (g/mol) 286.67
IUPAC Name (2R,3R,4S,5R)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(Cl)ncnc23

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H11ClN4O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2/t4-,6-,7-,10-/m1/s1
Formula Weight 286.0469
Chemical Name or Material 6-Chloropurine-9-beta-D-ribofuranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.