missing translation for 'onlineSavingsMsg'
Learn More

6-chloropurine, 99+%

Product Code. 10404775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10404775 25 g Glass bottle 25g
10174090 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10404775 Supplier Thermo Scientific Acros Supplier No. 171280250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 87-42-3 | C5H3ClN4 | 154.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 87-42-3
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.56
MDL Number MFCD00075825
InChI Key ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Synonym 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
PubChem CID 5359277
IUPAC Name 6-chloro-7H-purine
SMILES C1=NC2=C(N1)C(=NC=N2)Cl

Spécification

CAS Min % 99.0
CAS Max % 100.0
Melting Point >300°C
Color Orange-Yellow to Yellow
Elemental Analysis (on dry substance) :
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 2163
Solubility Information Solubility in water: 5g/L water
Formula Weight 154.56
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 6-chloropurine
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 6-Chloropurine
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.