missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine, 99%

Product Code. 11453514
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11453514

Brand: Thermo Scientific Alfa Aesar A11202.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-42-3 | C5H3ClN4 | 154.557 g/mol

6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

Solubility
Soluble in hot water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-42-3
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.557
MDL Number MFCD00075825
InChI Key ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Synonym 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
PubChem CID 5359277
IUPAC Name 6-chloro-7H-purine
SMILES C1=NC2=C(N1)C(=NC=N2)Cl

Specifications

Melting Point >300°C (decomposition)
Quantity 25 g
Beilstein 5774
Merck Index 14,2161
Solubility Information Soluble in hot water and dimethyl formamide.
Formula Weight 154.56
Percent Purity 99%
Chemical Name or Material 6-Chloropurine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.