missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine, MedChemExpress

Product Code. 16419416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419416 Supplier Medchem Express Supplier No. HYY0247
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Chloropurine is a building block in chemical synthesis. Intermediate in the preparation of 9-alkylpurines and 6-rnercaptopurine. Antitumor activities.

6-Chloropurine is a building block in chemical synthesis. Intermediate in the preparation of 9-alkylpurines and 6-rnercaptopurine. Antitumor activities.

Specifications

Chemical Name or Material 6-Chloropurine
CAS 87-42-3
Color White to Yellow
Purity Grade Notes Research
Molecular Formula C5H3ClN4
Quantity 100 mg
Synonym 6-Chloro-9H-purine
Solubility Information DMSO : 27.5 mg/mL (177.92 mM; Need ultrasonic) ∣H2O : 3 mg/mL (19.41 mM; Need ultrasonic)
SMILES ClC1=C2NC=NC2=NC=N1
Molecular Weight (g/mol) 154.56
Formula Weight 154.56
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.