missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyrimidine-2,4-diamine, 97%, Thermo Scientific™

Product Code. 10672255
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10672255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10672255 Supplier Thermo Scientific Maybridge Supplier No. BTB10268DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 156-83-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H5ClN4
MDL Number MFCD00006097
Quantity 1 g
Synonym 4-chloro-2,6-diaminopyrimidine, 2,4-diamino-6-chloropyrimidine, 2,6-diamino-4-chloropyrimidine, 6-chloro-2,4-diaminopyrimidine, 6-chloro-pyrimidine-2,4-diamine, 2,4-pyrimidinediamine, 6-chloro, 4-chloro-2,6-diamino pyrimidine, 2-amino-6-chloro-4-pyrimidinylamine, caap, minoxidil impurity b
InChI Key QJIUMVUZDYPQRT-UHFFFAOYSA-N
SMILES C1=C(N=C(N=C1Cl)N)N
IUPAC Name 6-chloropyrimidine-2,4-diamine
Molecular Weight (g/mol) 144.562
PubChem CID 67432
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 6-chloropyrimidine-2, 4-diamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.