missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorouracil, 98+%, Thermo Scientific™

Product Code. 11331427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331427 5 g 5g
11321427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331427 Supplier Thermo Scientific Alfa Aesar Supplier No. L01875.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chlorouracil (4-Chlorouracil; 6-Chlorouracil) is a halogenated uracil that is useful in studies of the effects of halogenation on nucleic acid base-pair stability and alkali metal ion affinity. Reaction of 6-chlorouracil with 4-(dimethylamino)pyridine, 4-methylpyridine, and pyridin-4-yl-morpholine yielded pyridinium-substituted uracils as chlorides which were converted into pyridinium uracilates by deprotonation. These heterocyclic mesomeric betaines are cross-conjugated and thus possess separate cationic (pyridinium) and anionic (uracilate) moieties. Calculations and X-ray single crystal analyses may be used to characterize these systems and to compare the salts with the betaines.

Solubility
Soluble in Ammonium Hydroxide

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4270-27-3
Molecular Formula C4H3ClN2O2
Molecular Weight (g/mol) 146.53
MDL Number MFCD00014595
InChI Key PKUFNWPSFCOSLU-UHFFFAOYSA-N
Synonym 6-chlorouracil, 6-chloropyrimidine-2,4 1h,3h-dione, 4-chlorouracil, 6-chloropyrimidine-2,4-diol, 6-chloro-2,4 1h,3h-pyrimidinedione, 6-chloro-2,4-dihydroxypyrimidine, 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-chloro-2,4-pyrimidinediol, 6-chloro-1,3-dihydropyrimidine-2,4-dione, 4-chloro-2,6-dihydroxypyrimidine
PubChem CID 73272
IUPAC Name 6-chloro-1H-pyrimidine-2,4-dione
SMILES ClC1=CC(=O)NC(=O)N1

Specifications

Melting Point ∽295°C (decomposition)
Quantity 5 g
Beilstein 120492
Solubility Information Soluble in Ammonium Hydroxide
Formula Weight 146.53
Percent Purity ≥98%
Chemical Name or Material 6-Chlorouracil
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.