missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorouracil, TRC

Product Code. 30365249
Change view
Click to view available options
Quantity:
50 mg
10 g
100 g
Unit Size:
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365249 100 g 100g
30351911 10 g -
30359191 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365249 Supplier Toronto Research Chemicals Supplier No. TRCC423780100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4270-27-3
Molecular Formula C4 H3 Cl N2 O2
Molecular Weight (g/mol) 146.53
Synonym 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloro-2,4-pyrimidinediol, 6-Chloropyrimidine-2,4(1H,3H)-dione, NSC 210419, NSC 43265
IUPAC Name 6-chloro-1H-pyrimidine-2,4-dione
SMILES ClC1=CC(=O)NC(=O)N1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H3ClN2O2/c5-2-1-3(8)7-4(9)6-2/h1H,(H2,6,7,8,9)
Formula Weight 145.9883
Chemical Name or Material 6-Chlorouracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.