Learn More
Invitrogen™ Mouse 6-Ckine (CCL21A) ELISA Kit
Description
The Mouse 6-Ckine (CCL21A) ELISA quantitates Ms 6-Ckine in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms 6-Ckine. Principle of the method The Mouse 6-Ckine solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Spezifikation
| Accession Number | P84444 |
| Assay Range | 31.25 to 1000 pg/mL |
| Assay Sensitivity | 20 pg/mL |
| Calibration Range | 20 pg/ml to 1,000 pg/ml |
| Description | 6-Ckine (CCL21A) ELISA Kit, 96 Assays, 1 Kit, Mouse Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of 6-Ckine (CCL21A) in Samples Using a Microplate Reader |
| Detectable Analytes | 6-Ckine (CCL21A) |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.