Learn More
BD Biosciences™ 6-color TBNK Reagent with BD Trucount™ Tubes (CE/IVD)
Description
- Six-color direct immunofluorescence reagent for use with BD FACSCanto™ and BD FACSCanto™ II flow cytometers to identify and determine the percentages and absolute counts of T, B, and natural killer (NK) cells as well as the CD4 and CD8 subpopulations of T cells in peripheral blood.
- BD Multitest 6-color TBNK reagent and BD Trucount tubes can be used with the BD FACS™ Loader.
- Contents: CD3 FITC / CD16 PE + CD56 PE / CD45 PerCP-Cy™5.5 / CD4 PE-Cy™7 / CD19 APC / CD8 APC-Cy™7 and BD Trucount™ tubes.
Specifications
Specifications
| Content And Storage | CD3 FITC / CD16 PE + CD56 PE / CD45 PerCP-Cy™5.5 / CD4 PE-Cy™7 / CD19 APC / CD8 APC-Cy™7 and BD Trucount™ tubes. |
| No. of Tests | 50 Tests |
| Quantity | 50 Tests |
| Stability | Use tubes within 1 hour after removal from the foil pouch. Use remaining tubes within 1 month after opening the pouch. |
| Product Type | Immunofluorescence Flow Cytometry Test Reagents |