missing translation for 'onlineSavingsMsg'
Learn More

6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol, TRC

Product Code. 30138110
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138110 2.5 g 2.5g
30133579 500 mg 500mg
30011275 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138110 Supplier Toronto Research Chemicals Supplier No. TRCD4627632.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29529-99-5
Molecular Formula C11H20N4S2
Molecular Weight (g/mol) 272.43
Synonym 1,3,5-Triazine-2,4(1H,3H)-dithione, 6-(dibutylamino)-, 6-(Dibutylamino)-1,3,5-triazine-2,4(1H,3H)-dithione (ACI), s-Triazine-2,4-dithiol, 6-(dibutylamino)- (6CI, 8CI), 2,4-Dimercapto-6-(dibutylamino)triazine, 2-(Dibutylamino)-1,3,5-triazine-4,6-dithiol, 2-(Dibutylamino)-4,6-dimercapto-1,3,5-triazine, 2-(Dibutylamino)-4,6-dimercapto-s-triazine, 2-(Dibutylamino)-4,6-dithiol-s-triazine, 2-(Dibutylamino)-s-triazine-4,6-dithiol, 2-(N,N-Dibutylamino)-4,6-dimercapto-1,3,5-triazine, 2-Di-n-butylamino-4,6-dimercapto-S-triazine, 2-Dibutylamino-4,6-dithio-s-triazine, 6-(Dibutylamino)-1,3,5-s-triazine-2,4-dithiol, 6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol, 6-(Dibutylamino)-s-triazine-2,4-dithiol, 6-Dibutylamino-2,4-dimercapto-1,3,5-triazine, 6-N,N-Dibutylamino-1,3,5-triazine-2,4-dithione, Actor BSH, DB, DB (thiol), DB-N, PC 10, PC 10 (crosslinking agent), Triazine DB, Zisnet DB, Zisnet DB/LDH, Zisnet MP
SMILES CCCCN(CCCC)c1nc(S)nc(S)n1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H20N4S2/c1-3-5-7-15(8-6-4-2)9-12-10(16)14-11(17)13-9/h3-8H2,1-2H3,(H2,12,13,14,16,17)
Formula Weight 272.1129
Chemical Name or Material 6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.