missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2-mercaptobenzothiazole, 99%

Product Code. 11341587
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11341587

Brand: Thermo Scientific Alfa Aesar L04447.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-53-6 | C9H9NOS2 | 211.297 g/mol

6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-53-6
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
MDL Number MFCD00005782
InChI Key HOASVNMVYBSLSU-UHFFFAOYSA-N
Synonym 6-ethoxy-2-mercaptobenzothiazole, 6-ethoxybenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-ethoxy, 6-ethoxybenzothiazolethiol, usaf pd-58, 6-ethoxy-2-benzothiazolethiol, 6-ethoxybenzo d thiazole-2 3h-thione, 6-ethoxy-1,3-benzothiazole-2-thiol, 2-mercapto-6-ethoxybenzothiazole, 2-benzothiazolethiol, 6-ethoxy
PubChem CID 719344
IUPAC Name 6-ethoxy-3H-1,3-benzothiazole-2-thione
SMILES CCOC1=CC2=C(C=C1)NC(=S)S2

Specifications

Melting Point 196°C to 201°C
Quantity 25 g
Beilstein 152313
Solubility Information Insoluble in water.
Formula Weight 211.32
Percent Purity 99%
Chemical Name or Material 6-Ethoxy-2-mercaptobenzothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.