missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole Hydrochloride, TRC

Product Code. 30137122
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137122 5 g 5g
30134394 1 g 1g
30008283 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137122 Supplier Toronto Research Chemicals Supplier No. TRCF5955805G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84163-13-3
Molecular Formula C12 H13 F N2 O . Cl H
Molecular Weight (g/mol) 256.7
Synonym 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole Hydrochloride (1:1), 1,2-Benzisoxazole, 6-fluoro-3-(4-piperidinyl)-, monohydrochloride (9CI), 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidine hydrochloride, 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole monohydrochloride, 6-Fluoro-3-(4-piperidinyl)benzisoxazole hydrochloride, 6-Fluoro-3-(piperidin-4-yl)benzo[d]isoxazole hydrochloride, 6-Fluoro-3-(piperidinyl-4-yl)-benzisoxazole hydrochloride
IUPAC Name 6-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
SMILES Cl.Fc1ccc2c(noc2c1)C3CCNCC3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H13FN2O.ClH/c13-9-1-2-10-11(7-9)16-15-12(10)8-3-5-14-6-4-8;/h1-2,7-8,14H,3-6H2;1H
Formula Weight 256.0779
Chemical Name or Material 6-fluoro-3-(piperidin-4-yl)benzoisoxazole hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.