missing translation for 'onlineSavingsMsg'
Learn More

(6-Fluoro-4h-1,3-benzodioxin-8-yl)methylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10157622
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157622 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157622 Supplier Thermo Scientific Maybridge Supplier No. CC01963DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 859833-12-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H11ClFNO2
MDL Number MFCD07772790
Quantity 1 g
Synonym 6-fluoro-4h-1,3-benzodioxin-8-yl methylamine hydrochloride, 6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl methanamine hydrochloride, 1-6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl methanamine hydrochloride, 6-fluoro-4h-1,3-benzodioxin-8-yl methanamine;hydrochloride, 6-fluoro-2h,4h-benzo e 1,3-dioxan-8-yl methylamine, chloride, 1-6-fluoro-2h,4h-1,3-benzodioxin-8-yl methanamine-hydrogen chloride 1/1
InChI Key FBHYQVQESWEYTE-UHFFFAOYSA-N
SMILES C1C2=CC(=CC(=C2OCO1)CN)F.Cl
IUPAC Name (6-fluoro-4H-1,3-benzodioxin-8-yl)methanamine;hydrochloride
Molecular Weight (g/mol) 219.64
PubChem CID 18525698
Formula Weight 219.64
Percent Purity 97%
Chemical Name or Material (6-Fluoro-4H-1, 3-benzodioxin-8-yl)methylamine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.