missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoronicotinic acid, 97%

Product Code. 15421308
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421308 250 mg 250mg
15411308 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421308 Supplier Thermo Scientific Alfa Aesar Supplier No. H55147.MD
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Alfa Aesar 6-Fluoronicotinic acid, 97%, Quantity: 1g, Melting Point: 144 deg.C to 148 deg.C, Molecular Weight: 141.101, Percent Purity: 97%, CAS: 403-45-2, Formula Weight: 141.1, Health Hazard 1: H315-H319-H335 | CAS: 403-45-2 | C6H4FNO2 | 141.10 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-45-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD01859863
InChI Key UJDLCTNVHJEBDG-UHFFFAOYSA-N
Synonym 6-fluoronicotinic acid, 2-fluoropyridine-5-carboxylic acid, 6-fluoro-3-pyridinecarboxylic acid, 2-fluoro-5-carboxypyridine, 2-fluoro-5-pyridinecarboxylic acid, 3-carboxy-6-fluoropyridine, 3-pyridinecarboxylic acid, 6-fluoro, 6-fluoro-nicotinic acid, 6-fluoronicotinc acid, 2-fluoro-5-pyridine carboxylic acid
PubChem CID 242819
IUPAC Name 6-fluoropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(F)N=C1

Specifications

Melting Point 144°C to 148°C
Quantity 250 mg
Formula Weight 141.1
Percent Purity 97%
Chemical Name or Material 6-Fluoronicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.