missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroveratraldehyde, 97+%

Product Code. 11464707
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464707 25 g 25g
11444707 1 g 1g
11454707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464707 Supplier Thermo Scientific Alfa Aesar Supplier No. B21202.14
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71924-62-4 | C9H9FO3 | 184.17 g/mol

6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71924-62-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00061108
InChI Key IBBYQNVXKFMSSI-UHFFFAOYSA-N
Synonym 6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
PubChem CID 603595
IUPAC Name 2-fluoro-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(F)=C(C=O)C=C1OC

Specifications

Melting Point 98°C to 100°C
Quantity 25 g
Beilstein 2109343
Sensitivity Air Sensitive
Formula Weight 184.17
Percent Purity ≥98%
Chemical Name or Material 6-Fluoroveratraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.