missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-methylthio-4(3H)-pyrimidinone, 97%, Thermo Scientific™

Product Code. 11412120
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412120 25 g 25g
11402120 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412120 Supplier Thermo Scientific Alfa Aesar Supplier No. H28708.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1979-98-2
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.18
MDL Number MFCD00006087
InChI Key AEXCUJUYEZIWJV-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-2-methylthiopyrimidine, 2-methylthio pyrimidine-4,6-diol, s-methylthiobarbituric acid, 4,6-dihydroxy-2-methylmercaptopyrimidine, 2-methylthiopyrimidine-4,6-diol, 4,6-dihydroxy-2-methythiopyrimidine, 2-methylsulfanyl pyrimidine-4,6-diol, 4,6-dihydroxy-2-methylthio pyrimidine, 2-methylthio-4,6-pyrimidinediol
PubChem CID 74795
IUPAC Name 4-hydroxy-2-methylsulfanyl-1H-pyrimidin-6-one
SMILES CSC1=NC(O)=CC(=O)N1

Specifications

Melting Point >300°C
Quantity 25 g
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-2-methylthio-4(3H)-pyrimidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.