missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxycaproic acid, 95%

Product Code. 10652952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10652952 1 g Glass bottle 1g
10525681 5 g Glass bottle 5g
13478518 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10652952 Supplier Thermo Scientific Acros Supplier No. 342240010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1191-25-9 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-25-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00046560
InChI Key IWHLYPDWHHPVAA-UHFFFAOYSA-N
Synonym 6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
PubChem CID 14490
ChEBI CHEBI:17869
IUPAC Name 6-hydroxyhexanoic acid
SMILES C(CCC(=O)O)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 38.0°C to 40.0°C
Color White
Density 0.9810g/mL
Boiling Point 113.0°C to 116.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula OH(CH2)5COOH
Refractive Index 1.44
Quantity 1 g
Specific Gravity 0.981
Formula Weight 132.16
Percent Purity 95%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material 6-Hydroxycaproic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxycaproic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.