missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyindole, 97%

Product Code. 10143712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143712 1 g Glass bottle 1g
10200163 5 g Glass bottle 5g
11966631 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143712 Supplier Thermo Scientific Acros Supplier No. 379010010
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2380-86-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-86-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key XAWPKHNOFIWWNZ-UHFFFAOYSA-N
Synonym 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
PubChem CID 524508
IUPAC Name 1H-indol-6-ol
SMILES C1=CC(=CC2=C1C=CN2)O

Specifications

Melting Point 124.0°C to 130.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 133.15
Percent Purity 97%
Physical Form Shiny Crystalline Powder
Chemical Name or Material 6-Hydroxyindole
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 6-Hydroxyindole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.