missing translation for 'onlineSavingsMsg'
Learn More

6-(Hydroxymethyl)benzothiazole, 97%, Thermo Scientific™

Product Code. 11966791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11966791 1 g 1g
11976791 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966791 Supplier Thermo Scientific Acros Supplier No. 441330010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 19989-66-3
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
InChI Key OZGXSRLIKDPNMX-UHFFFAOYSA-N
Synonym benzo d thiazol-6-ylmethanol, 6-hydroxymethyl benzothiazole, 6-benzothiazolemethanol, benzothiazol-6-yl-methanol, benzothiazol-6-yl methanol, benzothiazole-6-methanol, benzothiazol-6-ylmethanol, 6-hydroxymethylbenzothiazole, benzo d thiazol-6-yl-methanol
PubChem CID 17860385
IUPAC Name 1,3-benzothiazol-6-ylmethanol
SMILES C1=CC2=C(C=C1CO)SC=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 103.0°C to 105.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Formula Weight 165.22
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 6-(Hydroxymethyl)benzothiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.