missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1,4-benzodioxane, 95%, remainder mainly 5-isomer

Product Code. 11490651
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490651 25 g 25g
11480651 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490651 Supplier Thermo Scientific Alfa Aesar Supplier No. B20127.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57744-67-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57744-67-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00221460
InChI Key HZTMYTXWFHBHDC-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxyiodobenzene, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodobenzodioxane, acmc-1azse, 6-iodo-1,4-benzodioxane, 6-iodo-2h,3h-benzo e 1,4-dioxin, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 1,4-benzodioxin,2,3-dihydro-6-iodo
PubChem CID 2776176
IUPAC Name 6-iodo-2,3-dihydro-1,4-benzodioxine
SMILES IC1=CC=C2OCCOC2=C1

Specifications

Boiling Point 100°C to 102°C (0.25 mmHg)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 95%
Assay remainder mainly 5-isomer
Chemical Name or Material 6-Iodo-1,4-benzodioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.