missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1H-indazole, 97%, Thermo Scientific™

Product Code. 10220750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10220750 1 g Glass bottle 1g
10566450 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10220750 Supplier Thermo Scientific Acros Supplier No. 432420010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 261953-36-0
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.03
MDL Number MFCD04114695
InChI Key RSGAXJZKQDNFEP-UHFFFAOYSA-N
Synonym 6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
PubChem CID 12991241
IUPAC Name 6-iodo-1H-indazole
SMILES C1=CC2=C(C=C1I)NN=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208°C to 211°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 244.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Iodo-1H-indazole
Show More Show Less

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.