missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1H-indazole, 97%, Thermo Scientific™

Product Code. 10566450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10566450 5 g Glass bottle 5g
10220750 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10566450 Supplier Thermo Scientific Acros Supplier No. 432420050
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 261953-36-0
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.03
MDL Number MFCD04114695
InChI Key RSGAXJZKQDNFEP-UHFFFAOYSA-N
Synonym 6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
PubChem CID 12991241
IUPAC Name 6-iodo-1H-indazole
SMILES C1=CC2=C(C=C1I)NN=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208°C to 211°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 244.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Iodo-1H-indazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.