missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-2-methyl-1,3-benzothiazole, 90%, Thermo Scientific™

Product Code. 10193364
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193364 250 mg 250mg
10546324 1 g 1g
10103374 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193364 Supplier Thermo Scientific Maybridge Supplier No. CC54711CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 68867-20-9
Molecular Formula C8H6INS
Molecular Weight (g/mol) 275.107
InChI Key YVXGKAZJAUWMPM-UHFFFAOYSA-N
Synonym 6-iodo-2-methyl-benzothiazole, 6-iodo-2-methylbenzo d thiazole, 2-methyl-6-iodobenzothiazole, 6-iodo-2-methylbenzothiazole, benzothiazole, 6-iodo-2-methyl, benzothiazole,6-iodo-2-methyl
PubChem CID 807592
IUPAC Name 6-iodo-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=C(C=C2)I

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 275.11
Percent Purity 90%
Chemical Name or Material 6-Iodo-2-methyl-1, 3-benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.