missing translation for 'onlineSavingsMsg'
Learn More

6-Iodoquinazolin-4-one, TRC

Product Code. 30400017
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400017 25 g -
30391237 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400017 Supplier Toronto Research Chemicals Supplier No. TRCI71958025G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16064-08-7
Molecular Formula C8H5IN2O
Molecular Weight (g/mol) 272.05
Synonym 6-Iodo-4(3H)-quinazolinone, 6-Iodo-4(1H)-quinazolinone, 6-Iodo-4-quinazolinol, 6-Iodo-1H-quinazolin-4-one, 6-Iodo-3,4-dihydroquinazolin-4-one, 6-Iodo-3H-quinazolin-4-one, 6-Iodo-4(3H)-quinazolinone, 6-Iodo-4-quinazolone, 6-Iodoquinazolin-4-ol
SMILES Ic1ccc2NC=NC(=O)c2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H5IN2O/c9-5-1-2-7-6(3-5)8(12)11-4-10-7/h1-4H,(H,10,11,12)
Formula Weight 271.9447
Chemical Name or Material 6-Iodoquinazolin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.