missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 6-JOE, SE (6-Carboxy-4',5'-Dichloro-2',7'-Dimethoxyfluorescein, Succinimidyl Ester)

Product Code. 11550156
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11550156 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11550156 Supplier Invitrogen™ Supplier No. C6171MP
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

6-JOE, SE (6-Carboxy-4',5'-Dichloro-2',7'-Dimethoxyfluorescein, Succinimidyl Ester)

This single isomer preparation of the amine-reactive dye 6-JOE, SE has fluorescence characteristics that are red-shifted as compared to fluorescein (excitation/emission is 522/550 nm) and is one of the traditional fluorophores used in automated DNA sequencing.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type 6-JOE SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye JOE
Label Type Classic Dyes
Emission 550
Excitation 522

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.