missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine Monohydrate, TRC

Product Code. 30137115
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137115 2.5 g 2.5g
30135666 25 g 25g
18072750 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137115 Supplier Toronto Research Chemicals Supplier No. TRCM2254502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5 H4 N4 S . H2 O
Molecular Weight (g/mol) 170.19
Synonym 6H-Purine-6-thione, 1,2,3,9-tetrahydro-, hydrate (1:1), 6H-Purine-6-thione, 1,7-dihydro-, monohydrate (9CI), Purine-6-thiol, monohydrate (8CI), 6-Mercaptopurine monohydrate, 9H-Purine-6-thiol hydrate, Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate
IUPAC Name 7H-purine-6-thiol;hydrate
SMILES O.Sc1ncnc2nc[nH]c12

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H4N4S.H2O/c10-5-3-4(7-1-6-3)8-2-9-5;/h1-2H,(H2,6,7,8,9,10);1H2
Formula Weight 170.0262
Percent Purity >95%
Chemical Name or Material 6-Mercaptopurine monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.