missing translation for 'onlineSavingsMsg'
Learn More

6'-Methoxy-2'-acetonaphthone (Naproxen Impurity L), TRC

Product Code. 30133889
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133889 10 g 10g
17972075 1 g 1g
17940825 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133889 Supplier Toronto Research Chemicals Supplier No. TRCM26080510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3900-45-6
Molecular Formula C13 H12 O2
Molecular Weight (g/mol) 200.233
Synonym 1-(6-Methoxynaphthalen-2-yl)ethanone, Naproxen Imp. L (EP), Acetylnerolin
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanone
SMILES COc1ccc2cc(ccc2c1)C(=O)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H12O2/c1-9(14)10-3-4-12-8-13(15-2)6-5-11(12)7-10/h3-8H,1-2H3
Formula Weight 200.084
Percent Purity >95%
Chemical Name or Material Acetylnerolin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.