missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-methylbenzothiazole, 97%

Product Code. 15435147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435147 5 g 5g
15425147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435147 Supplier Thermo Scientific Alfa Aesar Supplier No. H32425.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2941-72-2 | C9H9NOS | 179.237 g/mol

6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2941-72-2
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00005795
InChI Key DYHLJSUORLPGNT-UHFFFAOYSA-N
Synonym 6-methoxy-2-methylbenzo d thiazole, 6-methoxy-2-methylbenzothiazole, benzothiazole, 6-methoxy-2-methyl, 2-methyl-6-methoxybenzothiazole, 6-methoxy-2-methyl-benzothiazole, benzothiazole,6-methoxy-2-methyl, 6-methoxy-2-methylbenzothiazol, #, benzothiazole, 6-methoxy-2-methyl-6ci,7ci,8ci,9ci
PubChem CID 76254
IUPAC Name 6-methoxy-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=C(C=C2)OC

Specifications

Density 1.204
Flash Point 109°C (228°F)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-methylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.